Learn More
Corning™ 384-Well Clear Polystyrene Microplates
GREENER_CHOICE
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 364-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
Brand:  Corning™ 3640
842.19 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Product Code. 10206051
  • 1181.76 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Standard
    Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
    Clear
    Polystyrene
    20 to 80 μL
    Non-treated
    Flat Bottom (F-well)
    100/Cs.
    Clear
    Without Lid
    16 x 24
    384-Well NBS™ Microplate
    384
    Non-sterile
    112 μL
    Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    25/Pk.