missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Nonbinding Surface (NBS™) Microplates

Product Code. 10728402 Shop All Corning Products
Click to view available options
Color:
Black
White
White, Clear Bottom
Well Color:
Black
Clear
White
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 10728402

Brand: Corning™ 3655

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhanced signal-to-noise ratio

  • Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
  • Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
  • Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity, and inhibits adhesion of a number of cell lines Stable to aqueous solutions containing EtOH
  • Use at temperature range of 4° to 37°C (39° to 99°F)
  • Flat-bottom wells
  • Compatible with a wide range of plate readers, liquid handlers, and robotic systems

Generic Bar Coded Plates

  • Code 128 bar code format ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified
  • Labels applied to all four sides of the plate No sequence number duplicates

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 16 x 24
Color Black
For Use With (Equipment) Plate readers, liquid handlers and robotic systems
Material Polystyrene
Volume (Metric) Well 112 μL
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Flat Bottom (F-well)
For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
Temperature Range (English) 39°F to 99°F
Temperature Range (Metric) 4°C to 37°C
Well Color Clear
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.