Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Optical Imaging Microplate
GREENER_CHOICE
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand:  Corning™ 3985BC
Product Code. 10001923
  • 2097.83 € / Pack of 100
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
0.06 cm2
Black, Clear Bottom
384
Sterile
112 μL
With generic barcode
20/Pk.
Clear
With Lid
16 x 24
Polystyrene
20 to 80 μL
Cell Culture
Flat Bottom (F-well)
100/Cs.
100/Cs.