missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Half Area High Content Imaging Glass Bottom Microplate
GREENER_CHOICE

High optical-quality glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems

Brand:  Corning™ 4580

Product Code. 13539050

  • 330.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
  • Half area 96-well microplate reduces reagent consumption.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Half-Area
8 x 12
Cycloolefin Copolymer
Sterile
10/Cs.
With Lid
Black
96
Provides a flat and optically clear surface that reduces autofocus time, increases throughput and is ideal for cell growth
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 96-Well Half Area High Content Imaging Glass Bottom Microplate > None

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.