Learn More
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates
GREENER_CHOICE
Optimized for high-throughput sitting-drop protein crystallization
710.00 € - 775.39 €
Specifications
Barcode Alphanumeric
Lid Without Lid
Array 8 x 12
Color Clear
No. of Wells 96
View More Specs
Products 4
Product Code Brand Well Shape Volume (Metric) Well Material Price Quantity & Availability  
Product Code Brand Well Shape Volume (Metric) Well Material Price Quantity & Availability  
10471963
GREENER_CHOICE
View accessory products View Documents
Corning™
3550
Conical Flat 1 μL Polymer
775.39 €
Pack of 50
Estimated Shipment: 22-05-2025
Add to basket
10057412
View accessory products View Documents
Corning™
3552
Round, Flat Bottom (F-well) 2 μL PZero This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10436593
View accessory products View Documents
Corning™
3553
Conical Flat 2 μL PZero
775.39 €
Pack of 50
Estimated Shipment: 24-07-2025
Add to basket
10327152
GREENER_CHOICE
View accessory products View Documents
Corning™
3556
Round 4 μL Cycloolefin Copolymer This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Corning™ 96-well and 384-well crystallization microplates are optimized for high throughput protein crystal growth and screening
  • Designed for sitting drop applications
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Multiple formats and versatility for custom options to maximize crystal formation, identification and analysis, and harvesting
  • - Choose from four unique protein well shapes
    - Options include one or three protein wells per reservoir well
  • Cyclic olefin copolymer (COC) offers strong chemical compatibility and good optical clarity
  • Reservoir numbers are embossed on each individual well for easy identification
Specifications
Specifications
Alphanumeric
8 x 12
96
Non-treated
10/Pk.
Without Lid
Clear
Non-sterile
50/Cs.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates