Learn More
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE
Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.
Brand:  Corning™ 3605
 View more versions of this product
Product Code. 10111822
  • 1064.77 € / Pack of 100
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Standard
    8 x 12
    Plate readers, liquid handlers and robotic systems
    96
    Non-sterile
    Round
    100/Cs.
    100/Cs.
    4° to 37°C
    Without Lid
    White
    Polystyrene
    75 to 200 μL
    Non-binding
    For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    25/Pk.
    39°F to 99°F
    White