missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
Shop All Corning Products

Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 50
Description
With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
- Unique design shields each well to minimize well-to-well cross-talk.
- You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 8 x 12 |
| Packaging | 10/Pk., 50/Cs. |
| Binding Type | Ultra-Low Attachment |
| Color | Clear |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Ultra-low Binding |
| Well Shape | Round Bottom (U-well) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction