missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
BIOPHARMAGREENER_CHOICE

Color:Black, Clear Bottom; Sterility:Sterile

Brand:  Corning™ 4515

Product Code. 15329740

  • 227.00 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.

  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Black, Clear Bottom
96
Sterile
300 μL
Closure
1/Pk.
With Lid
Ultra-Low Attachment
Polystyrene
0.075 to 0.30 mL
Ultra-low Binding
Round Bottom (U-well)
5/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number01125011 Certificate TypeCertificate of Quality Date02/04/2025 Catalog Number4515
Lot Number00925038 Certificate TypeCertificate of Quality Date22/03/2025 Catalog Number4515
Lot Number25524031 Certificate TypeCertificate of Quality Date01/02/2025 Catalog Number4515
Lot Number20724036 Certificate TypeCertificate of Quality Date29/01/2025 Catalog Number4515
Lot Number12924030 Certificate TypeCertificate of Quality Date25/06/2024 Catalog Number4515
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 96-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate > 1/Pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.