missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
GREENER_CHOICE

Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation

555.00 € - 2221.46 €

Specifications

Pore Size 3 μm
Type Microplate
Surface Coating BioCoat
Color Black, Clear
No. of Wells 96
View More Specs

Includes: A Falcon™ 24-Multiwell Insert Plate and a non-tissue culture-treated 24-well receiver plate with lid

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10737894
View Documents
Corning™
354149
1 N/A N/A N/A
10244152
GREENER_CHOICE
View Documents
Corning™
5 N/A N/A N/A
Description

Description

  • Highly reproducible assay system
  • Ideal for screening anti-angiogenic agents
  • Rapid image acquisition and data quantification
Specifications

Specifications

3 μm
BioCoat
96
Microplate
Black, Clear
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.