Learn More
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
GREENER_CHOICE
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand:  Corning™ 354541
 View more versions of this product
Product Code. 10526001
  • 486.88 € / Pack of 24
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Tested and found negative for bacteria and fungi
  • Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Order Info
Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12
3 μm
Adhesion, growth and differentiation of neuronal and transfected cell lines
2° to 8°C
PET
Well Plate
2
24