missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand: Corning™ 354469
90.86 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
- Coated with 0.2 micron filtered PDL in a cleanroom
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Poly-D-Lysine | |
Lid | |
100 mm |
Closure Included | |
Round | |
58.1 sq. cm | |
Round | |
Cell Culture Dish | |
10/cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction