missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-D-Lysine Culture Dish
GREENER_CHOICE

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Brand:  Corning™ 356468

427.62 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 10204281

  • 484.81 € / Pack of 100
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Coated with 0.2 micron filtered PDL in a cleanroom
Note: Dishes are not terminally sterilized and not assigned an SAL level. 

 

Order Info

Bulk quantities available; please contact your Customer Service Representative.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
Clear
Polystyrene
Poly-D-Lysine
Lid
60 mm
Closure Included
Round
21.3 sq. cm
Round
Cell Culture Dish
100/cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.