Learn More
Corning™ BioCoat™ Tumor Invasion Multiwell Plate
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Brand:  Corning™ 354166
 View more versions of this product
Product Code. 10007891
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running cell migration and invasion studies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.3 cm2
8 μm
5
5/Cs.
24
PET
Multiwell Plate
5/Pk.
Matrigel™ Matrix