Learn More
Corning™ Black 384-Well Polypropylene Assay Plate
Lower compound sticking and higher solvent resistance than polystyrene plates
846.89 €
Specifications
Format Standard
Array 16 x 24
Color Black
Material Polypropylene
No. of Wells 384
View More Specs
Products 1
Product Code Brand Lid Sterility Price Quantity & Availability  
Product Code Brand Lid Sterility Price Quantity & Availability  
10400672
View accessory products View Documents
Corning™
3658
Without Lid Non-sterile
846.89 €
Pack of 100
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Description
Description
  • Ideal for fluorescence assays and for compound storage
  • Round-bottom wells
  • Untreated
Specifications
Specifications
Standard
Black
384
16 x 24
Polypropylene
For fluorescence assays and compound storage