Learn More
Corning™ Black and White 96-Well Polypropylene Assay Plates
GREENER_CHOICE
Lower compound sticking and higher solvent resistance than polystyrene plates
Brand:  Corning™ 3356
507.80 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 10285421
  • 534.53 € / Pack of 100
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for fluorescence and luminescence assays and for compound storage
  • Round-bottom wells
  • Untreated surface
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
8 x 12
Polypropylene
75 to 200 μL
Non-treated
Ideal for fluorescence and luminescence assays and for compound storage
Without Lid
Black
96
Non-sterile
Round Bottom (U-well)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Black and White 96-Well Polypropylene Assay Plates > Black