missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Counter

The Corning Cell Counter is fast, accurate, low cost, and fits in the palm of your hand. With cloud-based cell counting, you always have the latest technology at your fingertips.

Brand:  Corning™ 6749

Product Code. 15813915

  • 4165.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For years, the choice between manual and automated cell counting has been a difficult one. Manual cell counting, on the one hand, is accurate, but time-consuming and very user-dependent. Automated cell counting, is much faster and less user-dependent, but the cost of disposable counting slides can be an issue. A tough choice, but now there is a solution.

The new Corning Cell Counter is the first automated cell counter that combines the best of both worlds. It is:

  • Fast – thanks to its online image processing
  • Accurate – thanks to its cloud-based machine learning algorithm.
  • Low-cost – works with common reusable glass hemocytometer. No consumables required.
  • Improved resolution – ability to accurately count as small as 4 µm (e.g., PBMC)

Services

1 Year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Counter
200X
USB 3.0
100 mm
90 mm
CE, usTUVcsa
Ability to accurately count as small as 4 µm (e.g., PBMC). Mammalian cells 4 to 70 μm.
Aluminum
10 μL
90 mm
1.0 kg
Cell Counting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.