Learn More
Corning™ Cell Culture Treated Flasks
BIOPHARMAGREENER_CHOICE
Manufactured from optically clear virgin polystyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Brand:  Corning™ 431080
 View more versions of this product
Product Code. 10296861
  • 395.00 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Stackable, polystyrene
  • Cell culture surface treatment promotes cell attachment and growth
  • Tested for sterility, toxicity, cell attachment, and growth promotion
  • Optically clear surfaces provide distortion-free microscope viewing
  • Printed lot number for product traceability
  • Sterile and nonpyrogenic
Cap Types
  • Phenolic-style screw caps for cultures requiring gas exchange
  • Plug seal caps for airtight cultures
  • Vent caps to minimize cell culture contamination with gas exchange
Canted-neck flasks feature:
  • Antitip skirt
  • Molded graduations
  • Frosted marking areas on both sides
  • Support bar across the top
  • Redesigned neck on 175cm2 flask features redesigned neck to aid media pouring, 1.4 in. (3.6cm) height for lower stacking height; vent cap has Costar™-style concentric circle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25.3 oz.
HDPE
Polystyrene
Lot numbers for ease in traceability
Rectangular
Yes
Vented
Orange
750 mL
For optimal cell attachment
Angled
Tissue Culture Treated
Sterile