missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell-Tak Cell and Tissue Adhesive
BIOPHARMAGREENER_CHOICE

Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.

Brand:  Corning™ 354241

1197.23 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 11583550

  • 1260.24 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stable for at least three months at 2° to 8°C. Do not freeze.
1 to 5μg/ cm2 of growth surface depending on cell type
Cell and Tissue Adhesive
5 mg
Molecular weight: 110 to 114kd
Liquid
As liquid in 5% acetic acid
Polyphenolic Proteins secreted by Mytilus edulis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.