missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell-Tak Cell and Tissue Adhesive

Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.

Brand:  Corning™ 354242

 View more versions of this product

Product Code. 11593550

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stable for at least three months at 2° to 8°C. Do not freeze.
1 to 5μg/ cm2 of growth surface depending on cell type
Cell and Tissue Adhesive
10 mg
Molecular weight: 110 to 114kd
Liquid
As liquid in 5% acetic acid
Polyphenolic Proteins secreted by Mytilus edulis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers