missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CellBIND™ Surface Culture Dishes
Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.
96.40 € - 238.00 €
Specifications
Closure Color | Clear |
---|---|
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Round |
Color | Clear |
Product Code | Brand | Diameter (Metric) | Culture Area | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Culture Area | Height (Metric) | Price | Quantity & Availability | |||
10581873
|
Corning™
3296 |
100 mm | 55 cm2 | 20 mm | N/A | ||||
10757804
|
Corning™
3294 |
35 mm | 8 cm2 | 10 mm |
238.00 €
Pack of 210 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
10665893
|
Corning™
3295 |
60 mm | 21 cm2 | 15 mm |
238.00 €
Pack of 126 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
Description
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
- Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
- 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
- 245 mm square dishes offer 500 cm2 growth surface
- Stacking beads aid in handling
- Vents provide consistent gas exchange
- Manufactured from optically clear virgin polystyrene
- Sterile
- Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
Specifications
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Sterile | |
Cell Culture Petri Dish |
Yes | |
Round | |
Single-use | |
Round | |
CellBIND | |
Lid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title