Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
BIOPHARMA
With low endotoxin content.
Brand:  Corning™ 21-030-CM
181.51 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 15333631
  • 191.06 € / Pack of 6
Explore more special offers
Description
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time. This formulation contains calcium and magnesium.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calcium and Magnesium
1 X
6 x 1000 mL
6.9 to 7.1
2° to 30°C
DPBS
Calcium Chloride 0.10g/L, Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Magnesium Chloride Hexahydrate 0.1g/L, Sodium Chloride 8g/L, Disodium Phosphate Heptahydrate 2.1716g/L