missing translation for 'onlineSavingsMsg'
Learn More

Corning™ G418 Sulfate, 50mg/mL
BIOPHARMA

Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

210.00 € - 554.88 €

Specifications

Content And Storage 2° to 8°C
Form Liquid
Product Type Antibiotic
Sterility Sterile
Format Plastic Bottle
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15303681
BIOPHARMA
View Documents
Corning™
30-234-CR
20 mL
210.00 €
20mL
Please to purchase this item. Need a web account? Register with us today!
15393671
BIOPHARMA
View Documents
Corning™
30-234-CI
100 mL
554.88 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Well-known, effective selection agent G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 μg/mL. This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene.  G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.
Specifications

Specifications

2° to 8°C
Antibiotic
Plastic Bottle
Liquid
Sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ G418 Sulfate, 50mg/mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.