missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Insulin-Transferrin-Selenium (ITS)

Click to view available options
Quantity:
10 mL
missing translation for 'unitSize'
10mL
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- 100X solution
- Animal free formulation
- Contains 1000mg/L human recombinant insulin, 550mg/L human recombinant transferrin, 0.67mg/L selenious acid
- Quantity: 10mL
Spécification
Spécification
| Content And Storage | 2° to 8°C |
| Form | Liquid |
| Product Type | ITS (Insulin, Transferrin, Selenium) |
| Sterility | Sterile-filtered |
| Format | Plastic Bottle |
| pH | 3 ±0.4 |
| Quantity | 10 mL |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu