missing translation for 'onlineSavingsMsg'
Learn More

Corning™ L-glutamine Solution
BIOPHARMA

Essential amino acid and a key component of culture media

115.14 € - 577.22 €

Specifications

Content And Storage -25° to -15°C
Form Liquid
Product Type L-Glutamine
Sterility Sterile-filtered
Format Plastic Bottle
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15393631
BIOPHARMA
View Documents
Corning™
25-005-CI
6 x 100 mL
115.14 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15303641
BIOPHARMA
View Documents
Corning™
25-005-CV
6 x 500 mL
577.22 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L-glutamine is an essential amino acid and a key component of culture media, serving as a major energy source for propagating cells. It is very stable as a dry powder and as a frozen solution but degrades rapidly in liquid media or stock solutions, producing toxic compounds. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use. This formulation is prepared in cell culture grade water.

  • Complete Certificate of Analysis available for each production lot, along with full formulation information
  • Produced under the highest industry standards to ensure superior results
Specifications

Specifications

-25° to -15°C
L-Glutamine
Plastic Bottle
Liquid
Sterile-filtered
5.5 ±0.5
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.