Learn More
Corning™ Metallocene Collection Container 2D Pillow Configuration
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Brand: Corning™ 91-100-30
This item is not returnable.
View return policy
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Metallocene | |
50 L | |
1 Ea. |
Sterile | |
Port 1, 2; 24 in. length tubing w/0.375 in. I.D. Male CPC conn. w/0.375 in. barb, dust cover and clamps; Port 3; 36 in. tubing 0.125 in. I.D. w/0.125 in. barb and male luer | |
1/Pk. |