missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Microbiological Media

Sterile filtered
240.31 € - 304.70 €
Specifications
Form | Liquid |
---|---|
Manufacturing Origin | United States |
Fill Volume | 6 x 1000 mL |
Formulation | Casein Peptone 10g/L, NaCl 10g/L |
For Use With (Application) | For maintenance/propagation of Escherichia coli with plasmid DNA and recombinant Protein production |
Product Code | Brand | Media Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Media Type | Price | Quantity & Availability | |||||
15343721
![]() |
Corning™
46-055-CM |
Terrific Broth |
304.70 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15333721
![]() |
Corning™
46-050-CM |
LB Broth |
240.31 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For the maintenance and propagation of E. coli
- Terrific Broth supports higher cell densities than LB Broth, increasing plasmid yield
Specifications
Liquid | |
6 x 1000 mL | |
For maintenance/propagation of Escherichia coli with plasmid DNA and recombinant Protein production | |
6 x 1000 mL |
United States | |
Casein Peptone 10g/L, NaCl 10g/L | |
Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Microbiological Media