missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sodium Pyruvate 100mM Solution
BIOPHARMA

Suggested replacement for 25-000-CIR

68.06 €

Specifications

Content And Storage 2° to 8°C
Form Liquid
Product Type Sodium Pyruvate
Sterility Sterile-filtered
Format Plastic Bottle
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15323581
BIOPHARMA
View Documents
Corning™
25-000-CI
100 mL
68.06 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

Specifications

Specifications

2° to 8°C
Sodium Pyruvate
Plastic Bottle
Liquid
Sterile-filtered
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Sodium Pyruvate 100mM Solution

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.