Learn More
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
BIOPHARMA
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.
79.65 € - 1314.15 €
Alert:
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15353651
BIOPHARMA
View Documents
Corning Media Tech
25-055-CI
6 x 100 mL Bottle
92.90 €
Pack of 6
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
15363651
BIOPHARMA
View Documents
Corning Media Tech
25-055-CV
6 x 500 mL Bottle
79.65 €
Pack of 6
In Stock
to see stock.
Add to basket
15373581
BIOPHARMA
View Documents
Corning Media Tech
25-055-CM
6 x 1 L Bottle
90.15 €
Pack of 6
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
15383651
BIOPHARMA
View Documents
Corning Media Tech
25-055-LB
20 L Bioprocess Container
372.93 €
20 L
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
15393651
BIOPHARMA
View Documents
Corning Media Tech
25-055-LG
1 x 100 L Bioprocess Container
919.95 €
100 L
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
15303661
BIOPHARMA
View Documents
Corning Media Tech
25-055-LH
1 x 200 L Bottle
1314.15 €
200 L
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Order Info
Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.