Learn More
Corning™ CellStripper Dissociation Reagent
BIOPHARMA
Non-enzymatic cell dissociation solution gently dislodges adherent cells in culture
Brand:  Corning™ 25-056-CI
100.64 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 15313661
  • 113.08 € / Pack of 6
Explore more special offers
Description
Description
Designed to be an alternative to trypsin when used in conjunction with serum-free or serum-containing media
  • Formulated with a proprietary mixture of chelators
  • Reduces the risk of cell damage associated with protein digestive enzymes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Cell Culture Dissociation Reagent
6 x 100 mL
Liquid
Sterile