Learn More
Corning™ Collagen I, Human
BIOPHARMAGREENER_CHOICE
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Brand:  Corning™ 354265
 View more versions of this product
Product Code. 13416809
  • 376.00 € / 10mg
Explore more special offers
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.2 to 2 μg/cm2
Frozen in 2 mm hydrogen chloride
Human Placenta
-20°C
Cell Attachment and Proliferation, Cell-specific Morphology and Function
10 mg
Collagen, Human Type I