Learn More
Corning™ Collagen I, Rat
BIOPHARMA
Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.
Brand:  Corning™ 356236
 View more versions of this product
Product Code. 10727531
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 to 4 mg/mL
Liquid in 0.02N acetic acid
Rat Tail Tendon
2°C to 8°C
Cell Attachment and Proliferation, Cell-specific Morphology and Function
1000 mg
Collagen, Rat Tail Type I
>90% by SDS-PAGE