Learn More
Corning™ Collagen IV, Human
GREENER_CHOICE
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Brand:  Corning™ 354245
Product Code. 11523560
  • 539.00 € / 250µg
Explore more special offers
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 5 μg/cm2
Frozen in 100 mm borate buffer pH 8.0
0.25 mg
Collagen, Human Type IV
Cell Attachment and Proliferation
8
Human Placenta
-70°C