Learn More
Corning™ Collagen IV, Mouse
GREENER_CHOICE
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Brand:  Corning™ 354233
 View more versions of this product
Product Code. 10376931
  • 178.00 € / 1mg
Explore more special offers
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 5 μg/cm2
Frozen in 0.05 M Hydrochloric Acid (HCL)
Collagen, Mouse Type IV
Cell Attachment and Proliferation
1 mg
-70°C