Learn More
Corning™ Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
GREENER_CHOICE
For use with 96-well plates
Brand:  Corning™ 4876
 View more versions of this product
Product Code. 10143811
  • 207.49 € / Pack of 100
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plastic
Open Reservoir Liner
100/Cs.
96
Sterile
Ideal for repetitively filling multi-channel pipettors
1/Pk.