Learn More
Corning™ Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
For use with 96-well plates
Brand: Corning™ 4878
180.95 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.Specifications
White | |
Sterile | |
Ideal for repetitively filling multi-channel pipettors | |
1/Pk. |
Polystyrene | |
Reservoir | |
100/Cs. | |
8 |