Learn More
Corning™ CrystalEX™ 96-Well Protein Crystallization Microplates
Designed for sitting-drop vapor diffusion high throughput protein crystallization
Brand: Corning™ 3773
Description
96-well Microplates
- Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
- Conical bottom protein wells allow improved centering of the protein drop.
- Compatible with manual pipettors and automation
- Novel merged well design provides efficient vapor space for protein crystallization.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
- Meets ANSI/SBS standard footprint dimensions for microplates
- Ideal for fully automated crystal screening
- Features 192 reservoir wells and 192 corresponding protein wells
- Flat bottom protein wells are optimized for imaging of crystals.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
Specifications
Standard | |
8 x 12 | |
96 | |
Non-treated | |
96 flat-bottom reservoir wells and 96 conical-bottom protein wells | |
10/Pk. | |
3.1 mm |
Without Lid | |
Clear | |
Non-sterile | |
Round | |
50/Cs. | |
50/Cs. |
Product Suggestions
Customers who viewed this item also viewed