Learn More
Corning™ DeckWorks™ Bulk Pipet Tips
Compatible with DeckWorks racks and packaged in resealable polyethylene bags
Brand:  Corning™ 4112
 View more versions of this product
Product Code. 11921841
  • 28.95 € / Pack of 1000
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer ­maximum savings.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 200 μL
Yellow
Round
Single-use
Yes
Polypropylene
Non-sterile
Help reduce the production and waste of plastic materials
Bulk
DeckWorks™ Racks
DeckWorks