Learn More
Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Nonsterile
Designed for quantitative assays in which sample binding can significantly alter results.
Brand:  Corning™ 4149
 View more versions of this product
Product Code. 12999577
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 300 μL
Natural
Hinged Rack
Hinged rack
Yes
Polypropylene
Non-sterile
Designed for quantitative assays in which sample binding can significantly alter results
Low Retention, Round
Universal Fit
DeckWorks