Learn More
Corning™ DeckWorks™ Reload Systems with Standard Tips
Product Code. 12939567 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
1 to 300 μL
100 to 1000 μL
Color:
Natural
Yellow
Unit Size:
Pack of 3072
Pack of 3840
Pack of 768
Pack of 960
Product Code. 12939567
Brand: Corning™ 4125
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Packaged in transparent trays for easy tip identification. Corning™ Deckworks™ Reload Systems with Standard Tips provide convenient and economical reloading of durable hinged racks
Corning DeckWorks reload systems are available in 10 μL, 200 μL, 300 μL, and 1,000 μL sizes and allow for convenient and economical reloading of durable Corning DeckWorks hinged racks. DeckWorks reload decks are packaged in transparent trays for easy tip identification, requires no transfer devices, saves space, and minimizes packaging waste. Although the pipet tips are made from virgin medical-grade resins, the reload decks and rack bases are manufactured from 100% recycled polypropylene. Nonsterile decks and racks are steam autoclavable.
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips is eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins, and/or peptides.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 to 10 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Format Rack
Tip Style Round
Packaging Reload system
Graduated Yes
Product Line DeckWorks
Material Polypropylene