missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)

Product Code. p-96954 Shop All Corning Products
Click to view available options
Quantity:
1 μg
100 μg
Purity or Quality Grade:
>95% by SDS-PAGE and >80% by pH 9.9 PAGE
>98% by SDS-PAGE and pH 9.9 PAGE
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 11593600

Brand: Corning™ 356001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhance the proliferation of a wide variety of cell types

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.

Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Product Type Growth Factor
Format Natural
Formulation Lyophilized from ammonium acetate, carrier free, reconstitute in dH2O
For Use With (Application) A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research
Purity or Quality Grade >95% by SDS-PAGE and >80% by pH 9.9 PAGE
Quantity 1 μg
Reactivity Mouse
Source Male Mouse Submaxillary Glands
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.