missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Rack for Conical Bottom Centrifuge Tubes

For conical bottom tubes

Brand:  Corning™ 431355

 View more versions of this product

Product Code. 10393023

  • 33.73 € / Pack of 20
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These polystyrene foam racks are designed to hold 50 Corning 15 mL conical bottom plastic centrifuge tubes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Centrifuge Tube Rack
Tubes (15 mL), Tubes (17.5 mm)
50
Conical bottom centrifuge tubes
20/Pk.
Not Autoclavable
Polystyrene
Non-sterile
20/Cs.
17.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.