Learn More
Corning™ Lambda Plus 8 and 12-Channel Pipettors
GREENER_CHOICE
Designed for universal fit with all common brands of pipet tips
Brand:  Corning™ 4084
 View more versions of this product
Product Code. 11373625
  • 614.18 € / Each
Estimated Shipment: 16-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
Order Info
Accessory stand and rack available separately (Cat. Nos. 07-201-790 and -789).
Services
3-Year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±8 to ±2%
Autoclavable
12
Non-sterile
1 to 10 μL
3 Years
CE
Universal Pipette Tip
Lambda Plus
Pipette
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Lambda Plus 8 and 12-Channel Pipettors > 1-10 μL; 12; None