Learn More
Corning™ Low-Volume 384-Well Black Flat Bottom Polystyrene Non-Treated Microplate
GREENER_CHOICE
Ideal for miniaturized 384-well fluorescent homogeneous assays
Brand:  Corning™ 3821
Product Code. 10726843
  • 523.37 € / Pack of 50
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Polystyrene
0.005 to 0.04 mL
Non-treated
For miniaturized 384-well fluorescent homogeneous assays
10/Pk.
Without Lid
Black
384
Non-sterile
Flat Bottom (F-well)
50/Cs.
50/Cs.