Learn More
Corning™ Matrigel™ Matrix
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Brand: Corning™ 356234
323.00 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Alert:
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
-20°C | |
No LDEV | |
Matrigel | |
Engelbreth-Holm-Swarm Mouse Tumor |
Basement Membrane Matrix | |
Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice | |
5 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.