missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ GFR Basement Membrane Matrix
GREENER_CHOICE

Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.

Brand:  Corning™ 356231

589.00 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 11553620

  • 620.00 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer

This item is not returnable. View return policy

Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

-20°C
No Phenol Red, No LDEV
10 mL
GFR Basement Membrane Matrix
Suitable for applications that require a highly defined basement membrane preparation.
Engelbreth-Holm-Swarm Mouse Tumor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.