missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ NutriStem™ hPSC XF Medium
Supports long-term culture of human pluripotent stem cells
Brand: Corning™ 40-05-100-1B
This item is not returnable.
View return policy
Description
Corning NutriStem hPSC XF medium is a defined, xeno-free, serum-free medium designed to support the growth and expansion of human induced pluripotent stem (hiPS) and human embryonic stem (hES) cells in a feeder-free environment.Specifications
-25° to -10°C | |
Undifferentiated hES and hiPS cells | |
Liquid | |
HSA | |
100 mL |
hPSC XF Medium | |
Serum-free, Xeno-free, cGMP | |
Sterile | |
Intended Use: For Research or Further Manufacturing Use Only |