missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Osteo Assay Surface Microplates
Capable of supporting the functional properties of osteogenic cells. Corning™ Osteo Assay Surface delivers lot-to-lot consistency, translating to reliable and reproducible results in bone cell assays. The surface also offers a consistent and defined alternative to preparing dentine or bone slices, reducing the variability in assay systems and resulting in more predictable assay readouts.
460.67 € - 1475.75 €
Specifications
Lid | With Lid |
---|---|
Color | Clear |
Material | Polystyrene |
Sterility | Sterile |
Surface Treatment | Osteo Assay |
Product Code | Brand | dimFormat | Volume (Metric) Working | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimFormat | Volume (Metric) Working | Packaging | Price | Quantity & Availability | |||
11357791
|
Corning™
3987 |
380 to 570 μL | 1/Pk; 4/Cs | ||||||
11377791
|
Corning™
3988 |
75 to 200 μL | 1/Pk; 4/Cs | ||||||
11397791
|
Corning™
3989 |
75 to 200 μL | 1/Pk.; 2/Cs. | ||||||
Description
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Specifications
With Lid | |
Polystyrene | |
Osteo Assay | |
1/Pk. |
Clear | |
Sterile | |
Osteoblast and osteoclast differentiation and functional studies |