missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PET Storage Bottles, Square with Cap
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
127.02 € - 313.85 €
Specifications
Color | Orange |
---|---|
Diameter (English) Inner Neck | 1.4 in. |
Diameter (Metric) Inner Neck | 37 mm |
For Use With (Application) | Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage |
Material | PET |
Product Code | Brand | Capacity | Diameter (English) | Graduations | Height (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Diameter (English) | Graduations | Height (English) | Price | Quantity & Availability | ||
15242596
|
Corning™
431530 |
4.2 oz., 125 mL | 2 in. | 50 to 125 mL | 4.19 in. |
127.02 €
Pack of 24 |
In Stock
Log in to see stock. |
||
15138125
|
Corning™
431531 |
8.45 oz., 250 mL | 2.3 in. | 50 to 250 mL | 5.6 in. |
158.60 €
Pack of 24 |
In Stock
Log in to see stock. |
||
15188135
|
Corning™
431532 |
16.9 oz., 500 mL | 3 in. | 100 to 500 mL | 6.9 in. |
243.13 €
Pack of 24 |
In Stock
Log in to see stock. |
||
15118145
|
Corning™
431533 |
33.8 oz., 1000 mL | 3.6 in. | 100 to 1000 mL | 8.5 in. |
313.85 €
Pack of 24 |
In Stock
Log in to see stock. |
||
Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10−6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic
Specifications
Orange | |
37 mm | |
PET | |
12/Pk. | |
Sterile | |
Orange | |
Single-use | |
Square | |
GL 45 |
1.4 in. | |
Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage | |
24/Cs. | |
12 Pk. | |
GL 45 | |
ISO 9001:2008 | |
Yes | |
HDPE | |
Plug Seal |