missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Premium Fetal Bovine Serum



FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
695.99 € - 1080.00 €
Specifications
Certifications/Compliance | cGMP |
---|---|
Color | Caramel/Amber |
Content And Storage | -25° to -15°C |
Product Type | Fetal Bovine Serum |
Quantity | 500 mL |
Product Code | Brand | Product Type | Heat Inactivated | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Heat Inactivated | Price | Quantity & Availability | ||||
15393681
![]() ![]() |
Corning™
35-016-CV |
Fetal Bovine Serum | Yes |
1080.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15383681
![]() ![]() |
Corning™
35-015-CV |
Fetal Bovine Serum | No |
695.99 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Premium FBS is sourced from U.S. herds
Compliance
USDA Certified FBS is sourced from USDA-certified facilities (abbatoirs).
Specifications
cGMP | |
-25° to -15°C | |
500 mL | |
Triple-filtered, 0.1 μm | |
Premium | |
Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only. | |
7.8 ±0.8 | |
USDA Inspected |
Caramel/Amber | |
Fetal Bovine Serum | |
Sterile | |
Bovine Fetus | |
≤20EU/ mL | |
<30 mg/dL | |
30 to 50g/L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Premium Fetal Bovine Serum