Learn More
Corning™ Primaria™ Cell Culture Multiwell™ Plates
GREENER_CHOICE
Features a surface treatment that is a permanent, stable modification of the polystyrene surface incorporating a mixture of anionic and cationic functional groups. Corning™ Primaria™ Cell Culture Multiwell™ Plates feature a flat bottom with low evaporation lid.
Brand:  Corning™ 353846
Product Code. 10196990
  • 214.36 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Primaria™ Cell Culture Multiwell™ Plates > 6-well